Learn More
Alkali Scientific Standing Cell Culture Inserts
SDP
Catalog No. 503029300
Encompass_Preferred
Closely mimic desired in vivo environments ASIStanding Cell Culture Inserts provide improved attachment growth and differentiation of various cell types With their permeable properties inserts are convenient and effective tool for diverse research areas such as transportation and invasion assays Specially selected track-etched membranes of three different pore sizes (0 4 3 0 and 8 0 um) are utilized Track-etched membranes ensure precise pore size distribution and consistent pore density providing well-controlled experimental parameters Standing Insert Material Polycarbonate (PC) stain-free low background interference Applicable experiments invasion migration transportation drug uptake epithelial polarization chemotoxis co-culture toxicity studies and in vivo barrier modeling Distance from membrane to the bottom of the well 6well Insert 0 91 mm 24well Insert 0 85 mm Nominal membrane thickness 10 23 Frame material Polystyrene (PS) Packed in cell culture-treated cell culture plate
Catalog No. 50-302-9300 Supplier Alkali Scientific Supplier No. TCS2403
Closely mimic desired in vivo environments ASIStanding Cell Culture Inserts provide improved attachment growth and differentiation of various cell types With their permeable properties inserts are convenient and effective tool for diverse research areas such as transportation and invasion assays Specially selected track-etched membranes of three different pore sizes (0 4 3 0 and 8 0 um) are utilized Track-etched membranes ensure precise pore size distribution and consistent pore density providing well-controlled experimental parameters Standing Insert Material Polycarbonate (PC) stain-free low background interference Applicable experiments invasion migration transportation drug uptake epithelial polarization chemotoxis co-culture toxicity studies and in vivo barrier modeling Distance from membrane to the bottom of the well 6well Insert 0 91 mm 24well Insert 0 85 mm Nominal membrane thickness 10 23 Frame material Polystyrene (PS) Packed in cell culture-treated cell culture plate