Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Standing Height Class 100 Clean Room Vinyl Sit-Stand with Stationary Glides

Catalog No. 15003994 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Adobe White
Black
Bluebird
Borscht
Burgundy
Carbon
Charcoal
Colonial Blue
Dove
Elephant Gray
Graphite
Imperial Blue
Ink
Mandarin
Marine Blue
Orange Kist
Sterling
Storm
Taupe
Material:
Vinyl (Grade A)
Vinyl (Grade B)
Vinyl (Grade C)
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
15-003-994 Dove Vinyl (Grade B)
15-002-466 Mandarin Vinyl (Grade C)
15-002-542 Imperial Blue Vinyl (Grade B)
15-002-595 Bluebird Vinyl (Grade C)
15-002-861 Black Vinyl (Grade B)
15-002-862 Ink Vinyl (Grade C)
15-002-863 Colonial Blue Vinyl (Grade B)
15-002-864 Taupe Vinyl (Grade A)
15-003-115 Marine Blue Vinyl (Grade A)
15-003-116 Taupe Vinyl (Grade B)
15-003-117 Sterling Vinyl (Grade A)
15-003-192 Burgundy Vinyl (Grade B)
15-003-456 Borscht Vinyl (Grade A)
15-003-488 Adobe White Vinyl (Grade B)
15-003-489 Elephant Gray Vinyl (Grade C)
15-003-519 Charcoal Vinyl (Grade B)
15-003-896 Black Vinyl (Grade C)
15-003-922 Storm Vinyl (Grade A)
15-003-924 Carbon Vinyl (Grade A)
15-003-993 Graphite Vinyl (Grade C)
15-004-007 Orange Kist Vinyl (Grade B)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 15-003-994
Only null left
Add to Cart
Add to Cart

Vinyl standing height sit-stand.

  • Adjustable from 21 to 31 in. from the top of the seat to the floor
  • Features seat height adjustment
  • 8-degree forward and 3-degree backward locking seat tilt
  • Cast-aluminum base
  • Black non-marring bell glides (suitable for both hard and carpeted floors)
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room

Specifications

Product Type Controlled Environment Chair
With Armrest No
With Casters No
With Tilt Yes
With Backrest No
Footrest No
With Lumbar Support No
Color Dove
Autoclavable Not Autoclavable
Material Vinyl (Grade B)
Seating Type High Bench Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.