Learn More
Lumacyte LLC STANDRD FLUID KIT FOR RADIANCE
Catalog No. NC2869395
Encompass
  • Essential for cell stabilization during analysis
  • Facilitates accurate characterization of cellular properties
  • Supports real-time single-cell analysis
  • Ensures cell integrity and stability
  • Enables reproducible data acquisition
  • Compatible with LumaCyte Radiance system
  • Utilizes Laser Force Cytology™ technology
  • Applicable in cell therapy, gene therapy, and vaccine development
  • Helps preserve native biophysical and biochemical properties of cells
  • Aids in enhanced data quality and insights into cell health
Catalog No. NC2869395 Supplier Lumacyte LLC Supplier No. LFKSTB3001
  • Essential for cell stabilization during analysis
  • Facilitates accurate characterization of cellular properties
  • Supports real-time single-cell analysis
  • Ensures cell integrity and stability
  • Enables reproducible data acquisition
  • Compatible with LumaCyte Radiance system
  • Utilizes Laser Force Cytology™ technology
  • Applicable in cell therapy, gene therapy, and vaccine development
  • Helps preserve native biophysical and biochemical properties of cells
  • Aids in enhanced data quality and insights into cell health