Learn More
MSC Stanley Fatmax Structural Foam Toolbox, 23 in., Water Resistant, 1 ea.
Catalog No. 5000510848
Encompass
This heavy-duty 23 in. structural foam toolbox is designed to provide secure and durable storage for power tools, hand tools, and other medium-to-large items. Constructed from thick, durable structural foam, this unit features a water-resistant design to protect contents from environmental elements.
  • Water-resistant design with integrated water seals
  • Equipped with sturdy metal latches
  • Features a portable half-tray for organized storage with space for larger items
  • Includes a lid-integrated V-groove with a ruler to hold materials like pipes and lumber in place for cutting
  • Ergonomic carry handle designed for comfortable transport
  • Stackable design for efficient storage
  • Rated IP53 for water and dust resistance
  • Supports loads up to 65 lb
Catalog No. 50-005-10848 Supplier MSC Supplier No. 82493818
This heavy-duty 23 in. structural foam toolbox is designed to provide secure and durable storage for power tools, hand tools, and other medium-to-large items. Constructed from thick, durable structural foam, this unit features a water-resistant design to protect contents from environmental elements.
  • Water-resistant design with integrated water seals
  • Equipped with sturdy metal latches
  • Features a portable half-tray for organized storage with space for larger items
  • Includes a lid-integrated V-groove with a ruler to hold materials like pipes and lumber in place for cutting
  • Ergonomic carry handle designed for comfortable transport
  • Stackable design for efficient storage
  • Rated IP53 for water and dust resistance
  • Supports loads up to 65 lb