Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, 99%, (Crystalline), MP Biomedicals

Catalog No. ICN15015791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15015791 1 kg
ICN15015780 100 g
ICN15015783 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15015791 Supplier MP Biomedicals, Inc Supplier No. 0215015791
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name λ²-tin(2+) dichloride
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99%
Formula Weight 189.6
Chemical Name or Material Stannous Chloride, Anhydrous
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.