Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Anhydrous (Crystalline Powder or Flakes/Technical), Fisher Chemical™

Catalog No. T141100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T141-100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T141-100

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Tin (II) Chloride Anhydrous
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name λ²-tin(2+) dichloride
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Melting Point 246°C
Color White
pH 2
Physical Form Solid
Quantity 100 g
Formula Weight 189.62
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Density 3.950g/cm³
Chemical Name or Material Stannous Chloride Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.