Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Dihydrate (Crystalline/Technical), Fisher Chemical

Catalog No. T163500 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
T163-500 Poly Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T163-500

Specifications

Boiling Point 652°C
Melting Point 37°C
Color White
CAS 10025-69-1
Physical Form Solid
Quantity 500 g
Molecular Formula Cl2Sn · 2 H2O
MDL Number MFCD00149863
Synonym Tin (II) Chloride
Formula Weight 225.65
Grade Technical
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Stannous Chloride Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. May cause an allergic skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.