Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, MP Biomedicals™

Catalog No. MP215015790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215015790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215015790 Supplier MP Biomedicals, Inc Supplier No. 0215015790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stannous Chloride, MP Biomedicals is used as a reducing agent, as a reagent in analytical chemistry and as a metal catalyst in organic reactions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Tin (II) Chloride, Tin Dichloride, Tin Protochloride
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name λ²-tin(2+) dichloride
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C (Lit.)
Melting Point 246°C (Lit.)
Physical Form Crystalline Powder
Quantity 500 g
Density 7.3g/cm3 at 20°C (Lit.)
Solubility Information Soluble in water.
Formula Weight 189.6
Percent Purity ∼99%
Recommended Storage Room Temperature (15–30°C), desiccate, store under nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.