Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Sulfate (Crystalline/Laboratory), Fisher Chemical

Catalog No. T154500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T154-500 500 mL HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. T154-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 7488-55-3
Molecular Formula O4SSn
Molecular Weight (g/mol) 214.77
MDL Number MFCD00011246
InChI Key OBBXFSIWZVFYJR-UHFFFAOYSA-L
Synonym stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
PubChem CID 62643
IUPAC Name λ²-tin(2+) sulfate
SMILES [Sn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 360°C
Color White
pH 2.0
Physical Form Solid
Quantity 500 mL
Formula Weight 214.77
Grade Laboratory
Identification Pass Test
Packaging HDPE Bottle
Chemical Name or Material Stannous Sulfate
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.