Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Sulfate, Technical, Spectrum™ Chemical
SDP

Catalog No. 18614052 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-052 500 g Poly Pail
18-614-050 2.5 kg Poly Pail
18-614-051 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-614-052 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1550500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7488-55-3

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 500 g
Molecular Weight (g/mol) 214.77
Grade Technical
Packaging Poly Pail
Chemical Name or Material Stannous Sulfate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.