Learn More
Bioss Staph. Enterotoxin
SDP
Catalog No. 5000066789
Encompass_Preferred
Staph Enterotoxin Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66789 Supplier Bioss Supplier No. BSM72429M
Staph Enterotoxin Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide