Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vela Diagnostics Great Basin Staph ID/R Blood Culture Panel

Catalog No. 23052005
Change view
Click to view available options
Quantity:
10 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-052-005 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-052-005 Supplier Vela Diagnostics Supplier No. 300785
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quickly identifies bloodstream pathogens directly from positive blood cultures found to contain gram-positive cocci in clusters or singles, meaning patients with likely contaminants or methicillin susceptible pathogens can be de-escalated without waiting for the final culture ID and sensitivity.

Unlike conventional methods, this sample-to-result assay is run directly from patient specimen, saving hand-on time and speeding appropriate therapy for the patient – improving outcomes and saving money.

Specifications

Content And Storage 2°C to 8°C
Description Sample-to-result panel: staph Pathogens identification
Clia Complexity Moderate
Detectable Analytes S. aureus, Staphylococcus species, C. jejuni, C. coli, mecA gene
Sample Type Pos Blood Culture
Quantity 10 Assays
Sample Size 50 μL
Test Time ≈90 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.