Learn More
ATCC Serratia marcescens
Supplier:  ATCC 43297
Encompass
Depositors claim multidrug resistance to amikacin, aztreonam, carbenicillin, cefoperazone, cefotaxime, cefoxitin, chloramphenicol, ciprofloxacin, mezlocillin, piperacillin, tetracycline, and tobramycin (see reference). ATCC could confirm antibiotic resistance only to cefoxitin and tobramycin.
Catalog No. 50-238-1286
 Restricted Item