Learn More
Abcam Recombinant Staphylococcus aureus Protein A, 10MG
Supplier:  Abcam AB5295310MG
Encompass_Preferred
Recombinant Staphylococcus aureus Protein A is a Staphylococcus aureus subsp. aureus NCTC 8325 Fragment protein, in the 32 to 327 aa range, expressed in Escherichia coli, with >99% purity, < 0.1 EU/mg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: SAOUHSC_00069, spa, Immunoglobulin G-binding protein A, IgG-binding protein A, Staphylococcal protein A, SpA
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2302718