Learn More
ATCC Staphylococcus aureus; Strain 27-05
Supplier:  ATCC BAA1768
Encompass
Staphylococcus aureus strain 27-05 is a drug-resistant bacterium with applications in antimicrobial resistance research. This strain is methicillin resistant and confirmed to be pvl negative and SCCmec type IV.
Catalog No. 50-238-0847