Learn More
ATCC Staphylococcus aureus; Strain HFH-30008
Supplier:  ATCC BAA1687
Encompass
Staphylococcus aureus strain HFH-30008 was isolated from a human sputum sample at Henry Ford Hospital in 2003. This strain is resistant to methicillin (MRSA) and has applications in antimicrobial resistance research.
Catalog No. 50-237-9868
 Restricted Item