Learn More
ATCC Staphylococcus aureus; Strain HFH-30032
Catalog No. 502381594
Encompass
Staphylococcus aureus strain HFH-30032 is a methicillin-resistant (MRSA) bacterium that was isolated in 2003 in Detroit, Michigan, from a human wound.
Catalog No. 50-238-1594 Supplier ATCC Supplier No. BAA1688
Add to Cart
Add to Cart
Staphylococcus aureus strain HFH-30032 is a methicillin-resistant (MRSA) bacterium that was isolated in 2003 in Detroit, Michigan, from a human wound.