Learn More
ATCC Staphylococcus aureus; Strain M10/0148
Catalog No. 502384816
Encompass
Staphylococcus aureus strain M10/0148 is a methicillin-resistant bacterium that was isolated in 2010 from a 64-year-old female in-patient at an acute care hospital in Dublin. This strain is pvl negative and was confirmed by PCR to have the mecC gene. This product has applications for drug discovery and emerging infectious disease research.
Catalog No. 50-238-4816 Supplier ATCC Supplier No. BAA2313
Staphylococcus aureus strain M10/0148 is a methicillin-resistant bacterium that was isolated in 2010 from a 64-year-old female in-patient at an acute care hospital in Dublin. This strain is pvl negative and was confirmed by PCR to have the mecC gene. This product has applications for drug discovery and emerging infectious disease research.