Learn More
ATCC Staphylococcus aureus; Strain MW2
Catalog No. 502381860
Encompass
Staphylococcus aureus strain MW2 [HIP08270] is a whole-genome sequenced bacterial strain. This culture is methicillin-resistant (MRSA) and is confirmed to have the pvl gene.
Catalog No. 50-238-1860 Supplier ATCC Supplier No. BAA1707
Staphylococcus aureus strain MW2 [HIP08270] is a whole-genome sequenced bacterial strain. This culture is methicillin-resistant (MRSA) and is confirmed to have the pvl gene.