Learn More
ATCC Staphylococcus aureus subsp. aureus Rosenbach
Supplier:  ATCC 29213/DEL
Encompass
Quality control strain for API, BBL, bioMerieux Vitek, Micro-Media, MicroScan™, and Sensititre products. Standard strain for CLSI antimicrobial susceptibility testing.
Catalog No. 50-189-137FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.