Learn More
ATCC Staphylococcus aureus subsp. aureus Rosenbach
Supplier:  ATCC 25923/DEL
Encompass
Quality control strain for the CAMP test, assay of wood smoke concentrate, evaluation of Mueller-Hinton agar, examination of dairy products, media testing, CLSI disk diffusion, and for Abbott, and Autobac products.
Catalog No. 50-189-132FP