Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC BAA42
Encompass
Staphylococcus aureus subsp. aureus strain HDE288 is a methicillin-resistant bacterium that was isolated from the pneumology ward of a hospital in Lisbon, Portugal. This strain has applications in antimicrobial resistance research, drug development, and infectious disease research.
Catalog No. 50-238-1779