Learn More
ATCC Staphylococcus aureus; subsp. aureus
Catalog No. 502381847
Encompass
Staphylococcus aureus subsp. aureus strain Newman D2C [NCTC 10833] is a whole-genome sequenced bacterium that can be used in the measurement of fibrinogen and fibrin degradation products in serum. This culture is a clumping factor-positive variant.
Catalog No. 50-238-1847 Supplier ATCC Supplier No. 25904
Staphylococcus aureus subsp. aureus strain Newman D2C [NCTC 10833] is a whole-genome sequenced bacterium that can be used in the measurement of fibrinogen and fibrin degradation products in serum. This culture is a clumping factor-positive variant.