Learn More
ATCC Staphylococcus aureus; subsp. aureus; SA113
Catalog No. 502381838
Encompass
Staphylococcus aureus subsp. aureus strain SA113 was isolated from a corneal ulcer. It can be used as a transformation host in molecular biology research.
Catalog No. 50-238-1838 Supplier ATCC Supplier No. 35556
Staphylococcus aureus subsp. aureus strain SA113 was isolated from a corneal ulcer. It can be used as a transformation host in molecular biology research.