Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier: ATCC 10832
Staphylococcus aureus subsp. aureus strain Wood 46 is a whole-genome sequenced bacterium with applications in dermatological research. This strain is known to produce dermatoxin.