Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC BAA44
Encompass
Staphylococcus aureus subsp. aureus strain HPV107 is an Iberian clone of MRSA that was isolated from a hospital in Lisbon, Portugal. This drug-resistant bacterium is SCCmec Type I and pvl negative.
Catalog No. 50-238-2293