Learn More
ATCC Staphylococcus aureus; subsp. aureus; Strain B8-31
Supplier:  ATCC BAA2094
Encompass
Staphylococcus aureus subsp. aureus strain B8-31 is a Gram-positive, whole-genome sequenced clinical isolate. This strain is mecA positive and has applications in antimicrobial resistance research.
Catalog No. 50-238-2459