Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC 29737
Encompass
Staphylococcus aureus subsp. aureus strain FDA is used in sterility testing and susceptibility disc testing. This strain is recommended as a test organism for USP <81>.
Catalog No. 50-238-2294
 Restricted Item