Learn More
ATCC Staphylococcus aureus; subsp. aureus; Strain NYBK2464
Supplier:  ATCC BAA41
Encompass
Staphylococcus aureus subsp. aureus strain NYBK2464 is a methicillin-resistant bacterium that was isolated from a hospital in New York, United States. This strain has applications in antimicrobial resistance research, drug development, and infectious disease research.
Catalog No. 50-238-2787