Learn More
ATCC Staphylococcus aureus; subsp. aureus; Mu50
Catalog No. 502383471
Encompass
This strain is Methicillin resistant (MRSA). Resistant to Oxacillin and shows reduced Vancomycin susceptibility. Genome sequenced strain.
Catalog No. 50-238-3471 Supplier ATCC Supplier No. 700699
Add to Cart
Add to Cart
This strain is Methicillin resistant (MRSA). Resistant to Oxacillin and shows reduced Vancomycin susceptibility. Genome sequenced strain.