Learn More
ATCC Staphylococcus aureus; subsp. aureus; Strain 252
Catalog No. 502384321
Encompass
Staphylococcus aureus subsp. aureus strain MRSA252 is a hospital-acquired methicillin-resistant strain (MRSA) isolated in the United Kingdom. This whole-genome sequenced bacterial strain has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-4321 Supplier ATCC Supplier No. BAA1720
Staphylococcus aureus subsp. aureus strain MRSA252 is a hospital-acquired methicillin-resistant strain (MRSA) isolated in the United Kingdom. This whole-genome sequenced bacterial strain has applications in antimicrobial resistance research and drug development.