Learn More
ATCC Staphylococcus aureus; subsp. aureus; genomic DNA
Supplier:  ATCC 33591D5
Encompass
Genomic DNA from Staphylococcus aureus subsp. aureus strain 328 (ATCC 33591). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-237-9526