Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC 700787
Encompass
Staphylococcus aureus subsp. aureus strain 2947 was isolated from blood culture in Port Chester, New York. This strain is resistant to methicillin and is pvl negative.
Catalog No. 50-237-9702
 Restricted Item