Learn More
ATCC Staphylococcus aureus; subsp. aureus; phage 3c
Catalog No. 502381003
Encompass
Staphylococcus aureus subsp. aureus bacteriophage 3C is propagated in Staphylococcus aureus subsp. aureus strain CDC 3C (ATCC 27703). Growth of this bacteriophage requires that certain media are used at specific times; the handling procedure must be followed as written and media cannot be interchanged or substituted.
Catalog No. 50-238-1003 Supplier ATCC Supplier No. 27703B1
Staphylococcus aureus subsp. aureus bacteriophage 3C is propagated in Staphylococcus aureus subsp. aureus strain CDC 3C (ATCC 27703). Growth of this bacteriophage requires that certain media are used at specific times; the handling procedure must be followed as written and media cannot be interchanged or substituted.