Learn More
ATCC Staphylococcus aureus; subsp. aureus; Strain TCH1516
Catalog No. 502381862
Encompass
Staphylococcus aureus subsp. aureus strain TCH1516 [USA300-HOU-MR] was isolated from an adolescent patient with severe sepsis syndrome at Texas Children's Hospital. This whole-genome sequenced MRSA strain has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-1862 Supplier ATCC Supplier No. BAA1717
Staphylococcus aureus subsp. aureus strain TCH1516 [USA300-HOU-MR] was isolated from an adolescent patient with severe sepsis syndrome at Texas Children's Hospital. This whole-genome sequenced MRSA strain has applications in antimicrobial resistance research and drug development.