Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC 43300
Encompass
Staphylococcus aureus subsp. aureus strain F-182 is a clinical isolate from Kansas. This bacterial strain is pvl negative and SCCmec type II, and it is resistant to methicillin and oxacillin.
Catalog No. 50-238-2012