Learn More
ATCC Staphylococcus aureus; subsp. aureus; genomic DNA
Catalog No. 502383962
Encompass
Genomic DNA isolated from Staphylococcus aureus subsp. aureus strain Seattle 1945. This bacterial strain is also available as ATCC Catalog No. 25923™.
Catalog No. 50-238-3962 Supplier ATCC Supplier No. 25923D5
Genomic DNA isolated from Staphylococcus aureus subsp. aureus strain Seattle 1945. This bacterial strain is also available as ATCC Catalog No. 25923™.