Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC 29213
Encompass
Staphylococcus aureus subsp. aureus strain Wichita was isolated from a wound. This product can be used in susceptibility disc testing, quality control, drug discovery, and enteric research.
Catalog No. 50-238-3720