Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC 25923
Encompass
Staphylococcus aureus subsp. aureus strain Seattle 1945 is a whole-genome sequenced clinical isolate. This culture is used as a quality control strain for the CAMP test, assay of wood smoke concentrate, evaluation of Mueller-Hinton agar, examination of dairy products, media testing, CLSI disk diffusion, and for Abbott, and Autobac products.
Catalog No. 50-238-4641