Learn More
ATCC Staphylococcus aureus; subsp. aureus
Catalog No. 502382581
Encompass
Staphylococcus aureus subsp. aureus strain 1063 was isolated from blood. This bacterial strain is resistant to gentamicin and methicillin and has applications in antimicrobial resistance research, drug development, and infectious disease research.
Catalog No. 50-238-2581 Supplier ATCC Supplier No. 33592
Staphylococcus aureus subsp. aureus strain 1063 was isolated from blood. This bacterial strain is resistant to gentamicin and methicillin and has applications in antimicrobial resistance research, drug development, and infectious disease research.