Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier: ATCC 33591
Staphylococcus aureus subsp. aureus strain 328 is whole-genome sequenced bacterium with applications in testing antimicrobial handwashing formulations and drug discovery. This strain is pvl negative and SCCmec type III, and it demonstrates resistance to methicillin.