Learn More
ATCC Staphylococcus capitis; subsp. capitis; Strain PRA 360 677
Catalog No. 502382630
Encompass
Staphylococcus capitis subsp. capitis strain PRA 360 677 is a whole-genome sequenced strain that can be used as a quality control strain for API products.
Catalog No. 50-238-2630 Supplier ATCC Supplier No. 35661
Staphylococcus capitis subsp. capitis strain PRA 360 677 is a whole-genome sequenced strain that can be used as a quality control strain for API products.