Learn More
ATCC Staphylococcus warneri
Supplier:  ATCC 27837
Encompass
Staphylococcus warneri strain RM 130 was isolated from human skin. This strain is grown aerobically on nutrient agar.
Catalog No. 50-238-0208