Learn More
ATCC Staphylococcus epidermidis (Winslow and Winslow) Evans
Supplier:  ATCC 12228/DEL
Encompass
Used in media testing and various assays. Organism used in genome sequencing project. This item is a QC bug for the following bioMérieux VITEK 2 Compact cards: BCL,NH, YST.
Catalog No. 50-189-129FP