Learn More
ATCC Treponema medium
Catalog No. 502380210
Encompass
Treponema medium strain G7201 is a bacterial type strain that was isolated in 1972 from the subgingival plaque of an adult male with periodontitis.
Catalog No. 50-238-0210 Supplier ATCC Supplier No. 700293
Treponema medium strain G7201 is a bacterial type strain that was isolated in 1972 from the subgingival plaque of an adult male with periodontitis.