Learn More
ATCC Staphylococcus haemolyticus
Supplier:  ATCC 700564
Encompass
Resistant to ampicillin, piperacillin-tazobactam, cefotaxime, cefazolin, and amoxicillin-clavulanate (interpretation of results is based on the product insert for the antibiotic). Resistant to ciprofloxacin, oxacillin, penicillin, clindamycin, erythromycin, ofloxacin, and gentamicin (interpretation of results is based on CLSI). Sensitive to tetracycline and vancomycin (interpretation of results is based on CLSI). Formerly Staphylococcus hominis.
Catalog No. 50-238-0712