Learn More
ATCC Staphylococcus epidermidis
Supplier: ATCC 14990
Staphylococcus epidermidis strain Fussel [NCTC 11047, R. Hugh 2466] is a whole genome-sequenced type strain that was isolated from the nose. This bacterium has applications in efficacy testing, media testing, inhibition testing, and quality control.