Learn More
ATCC Staphylococcus epidermidis; genomic DNA
Catalog No. 502381793
Encompass
Genomic DNA isolated from Staphylococcus epidermidis FDA strain PCI (ATCC 12228). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-1793 Supplier ATCC Supplier No. 12228D5
Genomic DNA isolated from Staphylococcus epidermidis FDA strain PCI (ATCC 12228). This whole-genome sequenced product can be used in PCR and other molecular biology applications.