Learn More
ATCC Staphylococcus epidermidis; Quantitative DNA
Catalog No. 502383584
Encompass
Quantitative genomic DNA from Staphylococcus aureus FDA strain PCI 1200 (ATCC 12228) can be used for assay development, verification, validation, and monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.
Catalog No. 50-238-3584 Supplier ATCC Supplier No. 12228DQ
Quantitative genomic DNA from Staphylococcus aureus FDA strain PCI 1200 (ATCC 12228) can be used for assay development, verification, validation, and monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.