Learn More
ATCC Staphylococcus epidermidis; Strain: AUS26
Catalog No. 502382591
Encompass
Staphylococcus epidermidis strain AUS26 is a multidrug-resistant bacterium that was isolated in 2017 from human blood in Australia. The depositor states that this strain is ST2-mixed lineage and is glycopeptide heteroresistant.
Catalog No. 50-238-2591 Supplier ATCC Supplier No. BAA3171
Staphylococcus epidermidis strain AUS26 is a multidrug-resistant bacterium that was isolated in 2017 from human blood in Australia. The depositor states that this strain is ST2-mixed lineage and is glycopeptide heteroresistant.